Skip to Content
Merck
2,2,6,6-Tetramethyl-4-piperidone hydrochloride 98%

2,2,6,6-Tetramethyl-4-piperidone hydrochloride

2,2,6,6-Tetramethyl-4-piperidone hydrochloride 98%
Empirical Formula (Hill Notation):
C9H17NO · HCl
CAS Number:
Molecular Weight:
191.70
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing