Skip to Content
Merck
(1S,2S)-1-Amino-2-benzyloxycyclopentane 98%

(1S,2S)-1-Amino-2-benzyloxycyclopentane

(1S,2S)-1-Amino-2-benzyloxycyclopentane 98%
Synonym(s):
(1S)-trans-2-(Phenylmethoxy)cyclopentanamine, (1S,2S)-trans-2-Benzyloxycyclopentylamine
Empirical Formula (Hill Notation):
C12H17NO
CAS Number:
Molecular Weight:
191.27

Products

Product Category
Brand
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing