Skip to Content
Merck
(1R,2S)-trans-2-Phenyl-1-cyclohexanol 99%

(1R,2S)-trans-2-Phenyl-1-cyclohexanol

(1R,2S)-trans-2-Phenyl-1-cyclohexanol 99%
Linear Formula:
C6H5C6H10OH
CAS Number:
Molecular Weight:
176.25

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing