Skip to Content
Merck
1-Boc-piperazine-2-carboxylic acid 95%

1-Boc-piperazine-2-carboxylic acid

1-Boc-piperazine-2-carboxylic acid 95%
Empirical Formula (Hill Notation):
C10H18N2O4
Molecular Weight:
230.26

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing