Skip to Content
Merck
1-Benzyl-3-piperidone hydrochloride hydrate 95%

1-Benzyl-3-piperidone hydrochloride hydrate

1-Benzyl-3-piperidone hydrochloride hydrate 95%
Empirical Formula (Hill Notation):
C12H15NO · HCl · xH2O
CAS Number:
Molecular Weight:
225.71 (anhydrous basis)
EC Number:

Products

Product Category
Brand
Formula Weight
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing