
(15:0)-Anacardic acid

Synonym(s):
Anacardic acid, 2-Hydroxy-6-pentadecylbenzoic acid, 22:0-Anacardic acid, 6-Pentadecylsalicylic acid
Empirical Formula (Hill Notation):
C22H36O3
CAS Number:
Molecular Weight:
348.52
Products
Brand
Formula Weight
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing