Skip to Content
Merck
1-(4-Nitrobenzenesulfonyl)-1H-1,2,4-triazole

1-(4-Nitrobenzenesulfonyl)-1H-1,2,4-triazole

1-(4-Nitrobenzenesulfonyl)-1H-1,2,4-triazole
Synonym(s):
1-(4-Nitrophenylsulfonyl)-1H-1,2,4-triazole, p-NBST
Empirical Formula (Hill Notation):
C8H6N4O4S
CAS Number:
Molecular Weight:
254.22
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing