Skip to Content
Merck
1,4-Dibromo-2,3-butanediol 95%, mixture of R,R and S,S isomers

1,4-Dibromo-2,3-butanediol

1,4-Dibromo-2,3-butanediol 95%, mixture of R,R and S,S isomers
Linear Formula:
BrCH2CH(OH)CH(OH)CH2Br
CAS Number:
Molecular Weight:
247.91

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing