Skip to Content
Merck
1,4-Cyclohexanedione mono(2,2-dimethyltrimethylene ketal) 95%

1,4-Cyclohexanedione mono(2,2-dimethyltrimethylene ketal)

1,4-Cyclohexanedione mono(2,2-dimethyltrimethylene ketal) 95%
Synonym(s):
3,3-Dimethyl-1,5-dioxaspiro[5.5]undecan-9-one
Empirical Formula (Hill Notation):
C11H18O3
CAS Number:
Molecular Weight:
198.26
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing