Skip to Content
Merck
1,3-Dibenzyloxy-2-propanol 97%

1,3-Dibenzyloxy-2-propanol

1,3-Dibenzyloxy-2-propanol 97%
Synonym(s):
1,3-Bis(benzyloxy)-2-propanol, 1,3-Di-O-benzylglycerol
Linear Formula:
(C6H5CH2OCH2)2CHOH
CAS Number:
Molecular Weight:
272.34

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing