Skip to Content
Merck
1,3-Bisbenzyl-2-oxoimidazolidine-4,5-dicarboxylic acid 97%

1,3-Bisbenzyl-2-oxoimidazolidine-4,5-dicarboxylic acid

1,3-Bisbenzyl-2-oxoimidazolidine-4,5-dicarboxylic acid 97%
Synonym(s):
2-Oxo-1,3-bis(phenylmethyl)-4,5-imidazolidinedicarboxylic acid
Empirical Formula (Hill Notation):
C19H18N2O5
CAS Number:
Molecular Weight:
354.36
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing