Skip to Content
Merck
1-(2-Methoxyethyl)piperazine 97%

1-(2-Methoxyethyl)piperazine

1-(2-Methoxyethyl)piperazine 97%
Synonym(s):
4-(2-Methoxyethyl)piperazine, N-(2-Methoxyethyl)piperazine
Empirical Formula (Hill Notation):
C7H16N2O
CAS Number:
Molecular Weight:
144.21

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing