Skip to Content
Merck
1-(2-Mesitylenesulfonyl)-3-nitro-1H-1,2,4-triazole 98%

1-(2-Mesitylenesulfonyl)-3-nitro-1H-1,2,4-triazole

1-(2-Mesitylenesulfonyl)-3-nitro-1H-1,2,4-triazole 98%
Synonym(s):
(2,4,6-Trimethylphenyl)-(3-nitro-1,2,4-triazol-1-yl) sulfone, MSNT
Empirical Formula (Hill Notation):
C11H12N4O4S
CAS Number:
Molecular Weight:
296.30

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing