Skip to Content
Merck
1-(2-Furoyl)piperazine 97%

1-(2-Furoyl)piperazine

1-(2-Furoyl)piperazine 97%
Synonym(s):
2-Furoic piperazide
Empirical Formula (Hill Notation):
C9H12N2O2
CAS Number:
Molecular Weight:
180.20
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing