Skip to Content
Merck
1,2-Dimyristoyl-rac-glycero-3-phosphocholine ≥99%

1,2-Dimyristoyl-rac-glycero-3-phosphocholine

1,2-Dimyristoyl-rac-glycero-3-phosphocholine ≥99%
Synonym(s):
1,2-Ditetradecanoyl-rac-glycerol-3-phosphocholine, rac-Phosphatidylcholine, 1,2-dimyristoyl, DL-α-Dimyristoyl-phosphatidylcholine, DL-β,γ-Dimyristoyl-α-lecithin
Empirical Formula (Hill Notation):
C36H72NO8P
CAS Number:
Molecular Weight:
677.93
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Functional Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing