
1,2-Dimethyl-3-propylimidazolium bis(trifluoromethylsulfonyl)imide

Synonym(s):
DMPIIm
Empirical Formula (Hill Notation):
C10H15F6N3O4S2
CAS Number:
Molecular Weight:
419.36
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Application
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing