1,1′-Difluoro-2,2′-bipyridinium bis(tetrafluoroborate)
Synonym(s):
MEC-31
Empirical Formula (Hill Notation):
C10H8B2F10N2
CAS Number:
Molecular Weight:
367.79
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing