Product Name
Dimethyl sulfoxide analytical standard
Product Number
94563
Product Brand
sial
CAS Number
67-68-5
Molecular Weight
78.13
Test
Specification
APPEARANCE (COLOR)Colorless
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 99.95 %
REFRACTIVE INDEX N20/D1.478 - 1.480
WATER (COULOMETR.)≤ 0.30 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS