Skip to Content
Merck
All Photos(1)

Key Documents

SAB2500967

Sigma-Aldrich

Anti-SNX16 antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-DKFZp666H147, Anti-Sorting nexin 16

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, bovine, canine, mouse, human

technique(s)

indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SNX16(64089)

General description

The gene SNX16 (sorting nexin 16) is mapped to human chromosome 8q21.13. It belongs to the sorting nexins (SNXs) family of proteins. The protein has a coiled coil domain and a PX domain, which helps in membrane association. It is present in early and late endosomes/lysosomes.

Immunogen

Peptide with sequence ATPYVPVPMPIGN-C from the N Terminus of the protein sequence according to NP_071416.2; NP_690049.1; NP_690050.1.

Biochem/physiol Actions

SNX16 (sorting nexin 16) participates in trafficking of proteins between the early and late endosomes. It is also involved in the delivery of viral (vesicular stomatitis virus) nucleocapsids from endosome to the cytosol. In a mouse model, SNX16 negatively regulates cell migration and tumorigenesis in breast cancer cells.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rong-Zong Liu et al.
Genomics, 92(6), 436-445 (2008-09-13)
We have identified a new member of the FABP gene family, designated FABP12. FABP12 has the same structure as other FABP genes and resides in a cluster with FABP4/5/8/9 within 300,000 bp chromosomal region. FABP12 orthologs are found in mammals
Leilei Zhang et al.
Cell regeneration (London, England), 2(1), 3-3 (2013-01-01)
Sorting nexins are a large family of proteins that are associated with various components of the endosome system and they play many roles in processes such as endocytosis, intracellular protein trafficking and cell signaling. The subcellular distribution patterns of many
Isabelle Le Blanc et al.
Nature cell biology, 7(7), 653-664 (2005-06-14)
During viral infection, fusion of the viral envelope with endosomal membranes and nucleocapsid release were thought to be concomitant events. We show here that for the vesicular stomatitis virus they occur sequentially, at two successive steps of the endocytic pathway.
Brendon J Hanson et al.
The Journal of biological chemistry, 278(36), 34617-34630 (2003-06-19)
Sorting nexins (SNXs) are a growing family of proteins characterized by the presence of a PX domain. The PX domain mediates membrane association by interaction with phosphoinositides. The SNXs are generally believed to participate in membrane trafficking, but information regarding

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service