Skip to Content
Merck
All Photos(1)

Key Documents

ATEH94724F89

AstaTech, Inc.

2,4-DIFLUORO-5-HYDROXYBENZONITRILE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
155.1
MDL number:

Pricing and availability is not currently available. Contact Technical Service

Iupac Name

2,4-difluoro-5-hydroxybenzonitrile

Inchi Code

1S/C7H3F2NO/c8-5-2-6(9)7(11)1-4(5)3-10/h1-2,11H

InChI key

ORAUQGDBWNRTOZ-UHFFFAOYSA-N

Purity

95%

Country of Origin

CN

Physical Form

Yellow to Brown Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=173245

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=C90825


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service