Skip to Content
Merck
All Photos(1)

Key Documents

178527

Sigma-Aldrich

N,N-Dimethylformamide dipropyl acetal

97%

Synonym(s):

1,1-Dipropoxy-N,N-dimethylmethylamine, 1,1-Dipropoxytrimethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NCH(OCH2CH2CH3)2
CAS Number:
Molecular Weight:
175.27
Beilstein:
1098524
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.409 (lit.)

bp

67-68 °C/14 mmHg (lit.)

density

0.854 g/mL at 25 °C (lit.)

SMILES string

CCCOC(OCCC)N(C)C

InChI

1S/C9H21NO2/c1-5-7-11-9(10(3)4)12-8-6-2/h9H,5-8H2,1-4H3

InChI key

NSLGQFIDCADTAS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N,N-Dimethylformamide dipropyl acetal was used in the quantification of cocaine and its metabolite, benzoyl ecgonine from urine matrix.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

114.8 °F - closed cup

Flash Point(C)

46 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N C Jain et al.
Journal of forensic sciences, 22(1), 7-16 (1977-01-01)
A gas chromatographic procedure has been developed for the simultaneous determination of cocaine and benzoyl ecgonine in urine specimens. The two drugs are extracted by isopropanol/chloroform from urine samples saturated with a bisalt buffer. The organic extract is evaporated to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service