Merck
  • Carbazole-azine based fluorescence 'off-on' sensor for selective detection of Cu
Carbazole-azine based fluorescence 'off-on' sensor for selective detection of Cu
Luminescence : the journal of biological and chemical luminescence (2017-06-21)
Denzil Britto Christopher Leslee, Sekar Karuppannan, Karmegam Muthu Vengaian, Sivaraman Gandhi, Singaravadivel Subramanian
ABSTRACT
A new carbazole-azine based fluorescent sensor was synthesized and characterized. The selectivity of the sensor for Cu
Sigma-Aldrich
9-Ethylcarbazole, 97%