Merck
  • The pharmacology of two novel long-acting phosphodiesterase 3/4 inhibitors, RPL554 [9,10-dimethoxy-2(2,4,6-trimethylphenylimino)-3-(n-carbamoyl-2-aminoethyl)-3,4,6,7-tetrahydro-2H-pyrimido[6,1-a]isoquinolin-4-one] and RPL565 [6,7-dihydro-2-(2,6-diisopropylphenoxy)-9,10-dimethoxy-4H-pyrimido[6,1-a]isoquinolin-4-one].
The pharmacology of two novel long-acting phosphodiesterase 3/4 inhibitors, RPL554 [9,10-dimethoxy-2(2,4,6-trimethylphenylimino)-3-(n-carbamoyl-2-aminoethyl)-3,4,6,7-tetrahydro-2H-pyrimido[6,1-a]isoquinolin-4-one] and RPL565 [6,7-dihydro-2-(2,6-diisopropylphenoxy)-9,10-dimethoxy-4H-pyrimido[6,1-a]isoquinolin-4-one].
The Journal of pharmacology and experimental therapeutics (2006-05-10)
Victoria Boswell-Smith, Domenico Spina, Alec W Oxford, Mike B Comer, Esther A Seeds, Clive P Page
ABSTRACT
The pharmacology of two novel, trequinsin-like PDE3/4 inhibitors, RPL554 [9,10-dimethoxy-2(2,4,6-trimethylphenylimino)-3-(N-carbamoyl-2-aminoethyl)-3,4,6,7-tetrahydro-2H-pyrimido-[6,1-a]isoquinolin-4-one] and RPL565 [6,7-dihydro-2-(2,6-diisopropylphenoxy)-9,10-dimethoxy-4H-pyrimido[6,1-a]isoquinolin-4-one], has been investigated in a number of in vitro and in vivo assays. Electrical field stimulation-induced contraction of guinea pig superfused isolated tracheal preparations was significantly inhibited by RPL554 (10 microM) and RPL565 (10 microM) (percentage control; 93 +/- 1.2 and 84.4 +/- 2.7, respectively). Contractile responses were suppressed for up to 12 h after termination of superfusion with RPL554 demonstrating a long duration of action. RPL554 and RPL565 inhibited, in a concentration-dependent manner, lipopolysaccharide-induced tumor necrosis factor alpha release from human monocytes [IC50; 0.52 microM (0.38-0.69) and 0.25 microM (0.18-0.35), respectively] and proliferation of human mononuclear cells to phytohemagglutinin [IC50; 0.46 microM (0.24-0.9) and 2.90 microM (1.6-5.4), respectively]. The inhibitory effect of these drugs in vitro was translated into anti-inflammatory activity in vivo. RPL554 (10 mg/kg) and RPL565 (10 mg/kg) administered orally significantly inhibited eosinophil recruitment following antigen challenge in ovalbumin-sensitized guinea pigs. Likewise, inhalation of dry powder containing RPL554 by conscious guinea pigs (25% in micronized lactose) 1.5 h before antigen exposure significantly inhibited the recruitment of eosinophils to the airways. Exposure of conscious guinea pigs to inhalation of dry powder containing RPL554 (2.5%) and RPL565 (25%) in micronized lactose significantly inhibited histamine-induced plasma protein extravasation in the trachea and histamine-induced bronchoconstriction over a 5.5-h period. Thus, RPL554 and RPL565 are novel, long-acting PDE 3/4 inhibitors exhibiting a broad range of both bronchoprotective and anti-inflammatory activities.
Sigma-Aldrich
RPL554, ≥98% (HPLC)
Sigma-Aldrich
Trequinsin hydrochloride, ≥98%