Merck
  • Protein Footprinting, Conformational Dynamics, and Core Interface-Adjacent Neutralization "Hotspots" in the SARS-CoV-2 Spike Protein Receptor Binding Domain/Human ACE2 Interaction.
Protein Footprinting, Conformational Dynamics, and Core Interface-Adjacent Neutralization "Hotspots" in the SARS-CoV-2 Spike Protein Receptor Binding Domain/Human ACE2 Interaction.
Journal of the American Society for Mass Spectrometry (2021-04-02)
Dominic Narang, D Andrew James, Matthew T Balmer, Derek J Wilson
ABSTRACT
The novel severe respiratory syndrome-like coronavirus (SARS-CoV-2) causes COVID-19 in humans and is responsible for one of the most destructive pandemics of the last century. At the root of SARS-CoV infection is the interaction between the viral spike protein and the human angiotensin converting enzyme 2 protein, which allows the virus to gain entry into host cells through endocytosis. In this work, we apply hydrogen-deuterium exchange mass spectrometry (HDX-MS) to provide a detailed view of the functional footprint and conformational dynamics associated with this interaction. Our results broadly agree with the binding interface derived from high resolution X-ray crystal structure data but also provide insights into shifts in structure and dynamics that accompany complexation, including some that occur immediately outside of the core binding interface. We propose that dampening of these "binding-site adjacent" dynamic shifts could represent a mechanism for neutralizing activity in a multitude of spike protein-targeted mAbs that have been found to specifically bind these "peripheral" sites. Our results highlight the unique capacity of HDX-MS to detect potential neutralization "hotspots" outside of the core binding interfaces defined by high resolution structural data.
Sigma-Aldrich
[Glu1]-Fibrinopeptide B human, ≥90% (HPLC)