Merck
  • SETDB1 promotes glioblastoma growth via CSF-1-dependent macrophage recruitment by activating the AKT/mTOR signaling pathway.
SETDB1 promotes glioblastoma growth via CSF-1-dependent macrophage recruitment by activating the AKT/mTOR signaling pathway.
Journal of experimental & clinical cancer research : CR (2020-10-17)
Shuai Han, Wei Zhen, Tongqi Guo, Jianjun Zou, Fuyong Li
ABSTRACT
Glioblastoma is a common disease of the central nervous system (CNS), with high morbidity and mortality. In the infiltrate in the tumor microenvironment, tumor-associated macrophages (TAMs) are abundant, which are important factors in glioblastoma progression. However, the exact details of TAMs in glioblastoma progression have yet to be determined. The clinical relevance of SET domain bifurcated 1 (SETDB1) was analyzed by immunohistochemistry, real-time PCR and Western blotting of glioblastoma tissues. SETDB1-induced cell proliferation, migration and invasion were investigated by CCK-8 assay, colony formation assay, wound healing and Transwell assay. The relationship between SETDB1 and colony stimulating factor 1 (CSF-1), as well as TAMs recruitment was examined by Western blotting, real-time PCR and syngeneic mouse model. Our findings showed that SETDB1 upregulated in glioblastoma and relative to poor progression. Gain and loss of function approaches showed the SETDB1 overexpression promotes cell proliferation, migration and invasion in glioblastoma cells. However, knockdown SETDB1 exerted opposite effects in vitro. Moreover, SETDB1 promotes AKT/mTOR-dependent CSF-1 induction and secretion, which leads to macrophage recruitment in the tumor, resulted in tumor growth. Our research clarified that SETDB1 regulates of tumor microenvironment and hence presents a potential therapeutic target for treating glioblastoma.
Sigma-Aldrich
Anti-SETDB1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.