
XTT sodium salt

Synonym(s):
2,3-Bis(2-methoxy-4-nitro-5-sulfophenyl)-2H-tetrazolium-5-carboxanilide inner salt
Empirical Formula (Hill Notation):
C22H16N7NaO13S2
CAS Number:
Molecular Weight:
673.52
Products
Brand
Formula Weight
Physical Form
Product Line
Purity
Detection Method
Quality Segment
Application
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing