
Prostaglandin B1

Synonym(s):
(13E,15S)-15-Hydroxy-9-oxoprosta-8(12),13-dien-1-oic acid, PGB1
Empirical Formula (Hill Notation):
C20H32O4
CAS Number:
Molecular Weight:
336.47
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing