
Naproxen Impurity I

Synonym(s):
2-(6-Methoxynaphthalen-2-yl)acetic acid, 6-Methoxy-2-naphthaleneacetic acid, 6-Methoxy-2-naphthylacetic acid
Empirical Formula (Hill Notation):
C13H12O3
CAS Number:
Molecular Weight:
216.23
Products
Brand
Formula Weight
Physical Form
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing