
Memantine Related Compound D

Synonym(s):
1-Bromo-3,5-dimethyladamantane, 1-Bromo-3,5-dimethyltricyclo[3.3.1.13,7]decane
Empirical Formula (Hill Notation):
C12H19Br
CAS Number:
Molecular Weight:
243.18
Products
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing