
Flonicamid metabolite TFNG

Synonym(s):
N-[[4-(Trifluoromethyl)-3-pyridinyl]carbonyl]glycine, 4-(Trifluoromethyl)nicotinoyl glycine
Empirical Formula (Hill Notation):
C9H7F3N2O3
CAS Number:
Molecular Weight:
248.16
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2