
Dicycloverine impurity A

Synonym(s):
[1,1′-Bicyclohexyl]-1-carboxylic acid
Empirical Formula (Hill Notation):
C13H22O2
CAS Number:
Molecular Weight:
210.31
Products
Formula Weight
Manufacturer
Format
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing