
Cucurbitacin E

Synonym(s):
α-Elaterin, α-Elaterine
Empirical Formula (Hill Notation):
C32H44O8
CAS Number:
Molecular Weight:
556.69
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Application
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing