Merck
  • A microbially derived tyrosine-sulfated peptide mimics a plant peptide hormone.
A microbially derived tyrosine-sulfated peptide mimics a plant peptide hormone.
The New phytologist (2017-05-31)
Rory N Pruitt, Anna Joe, Weiguo Zhang, Wei Feng, Valley Stewart, Benjamin Schwessinger, José R Dinneny, Pamela C Ronald
ABSTRACT
The biotrophic pathogen Xanthomonas oryzae pv. oryzae (Xoo) produces a sulfated peptide named RaxX, which shares similarity to peptides in the PSY (plant peptide containing sulfated tyrosine) family. We hypothesize that RaxX mimics the growth-stimulating activity of PSY peptides. Root length was measured in Arabidopsis and rice treated with synthetic RaxX peptides. We also used comparative genomic analyses and reactive oxygen species burst assays to evaluate the activity of RaxX and PSY peptides. Here we found that a synthetic sulfated RaxX derivative comprising 13 residues (RaxX13-sY), highly conserved between RaxX and PSY, induces root growth in Arabidopsis and rice in a manner similar to that triggered by PSY. We identified residues that are required for activation of immunity mediated by the rice XA21 receptor but that are not essential for root growth induced by PSY. Finally, we showed that a Xanthomonas strain lacking raxX is impaired in virulence. These findings suggest that RaxX serves as a molecular mimic of PSY peptides to facilitate Xoo infection and that XA21 has evolved the ability to recognize and respond specifically to the microbial form of the peptide.
Sigma-Aldrich
Phytagel, BioReagent, suitable for plant cell culture, powder
Sigma-Aldrich
Membrane Scaffold Protein 1E3D1 D73C, recombinant, expressed in E. coli, Cysteine substituted at position 73