Merck
  • STAT3 restrains RANK- and TLR4-mediated signalling by suppressing expression of the E2 ubiquitin-conjugating enzyme Ubc13.
STAT3 restrains RANK- and TLR4-mediated signalling by suppressing expression of the E2 ubiquitin-conjugating enzyme Ubc13.
Nature communications (2014-12-17)
Huiyuan Zhang, Hongbo Hu, Nathaniel Greeley, Jin Jin, Allison J Matthews, Erika Ohashi, Mauricio S Caetano, Haiyan S Li, Xuefeng Wu, Pijus K Mandal, John S McMurray, Seyed Javad Moghaddam, Shao-Cong Sun, Stephanie S Watowich
ABSTRACT
The transcriptional regulator STAT3 curbs pro-inflammatory cytokine production mediated by NF-κB signalling in innate immune cells, yet the mechanism by which this occurs has been unclear. Here we identify STAT3 as a pivotal negative regulator of Ubc13, an E2 ubiquitin-conjugating enzyme that facilitates TRAF6 K63-linked ubiquitination and NF-κB activation. Ubc13 accumulates intracellularly in the absence of STAT3. Depletion of Ubc13 in Stat3-deficient macrophages subdues excessive RANKL- or LPS-dependent gene expression, indicating that Ubc13 overexpression mediates enhanced transcriptional responses in the absence of STAT3. In RANKL-activated macrophages, STAT3 is stimulated by autocrine IL-6 and inhibits accrual of Ets-1, Set1 methyltransferase and trimethylation of histone H3 lysine 4 (H3K4me3) at the Ube2n (Ubc13) promoter. These results delineate a mechanism by which STAT3 operates as a transcriptional repressor on Ube2n, thus modulating NF-κB activity by regulation of Ubc13 abundance. Our data suggest that this pathway plays important roles in bone homeostasis and restraint of inflammation.
Sigma-Aldrich
Anti-Histone H3 Antibody, CT, pan, clone A3S, rabbit monoclonal, culture supernatant, clone A3S, Upstate®