Merck
  • Interactions between model triacylglycerol-rich lipoproteins and high-density lipoproteins in rat, rabbit and man.
Interactions between model triacylglycerol-rich lipoproteins and high-density lipoproteins in rat, rabbit and man.
Biochimica et biophysica acta (1991-02-05)
I J Martins, L Hopkins, C A Joll, T G Redgrave
ABSTRACT
There are inverse relationships between HDL cholesterol and plasma triacylglycerol concentrations in normal and in hypertriglyceridemic individuals. To investigate the interactions between triacylglycerol-rich lipid particles and HDL, a lipid emulsion model of the triacylglycerol-rich lipoproteins was prepared. When emulsion particles were incubated with rat high-density lipoproteins (HDL) in the presence of lipid transfer activity (d greater than 1.21 g/ml fractions) from rabbit or human plasma there was a rapid bi-directional exchange of cholesteryl oleate (CO) and phospholipid (PL) labels between lighter and heavier fractions of HDL and emulsion particles. The transfers of CO and PL labels between both light and heavy fractions of HDL and the emulsion particles were increased with increasing amounts of emulsion added to the incubations. Incubation with the d greater than 1.21 g/ml fraction from rat plasma resulted in only a small exchange of CO whereas PL exchange was similar to rabbit and human plasma. Retinyl palmitate label was not transferred from emulsion particles to the HDL fractions even in the presence of lipid transfer activity from rabbit or human plasma. The present study shows that the transfer protein-mediated exchanges of surface and core lipids between HDL and the triacylglycerol-rich lipoproteins are affected by the quantity of triacylglycerol-rich particles in the system. This mechanism may contribute to the inverse relationships between plasma triacylglycerol concentrations and HDL concentrations in normal and hypertriglyceridemic individuals.
Product Description
Supelco
Retinol palmitate, analytical standard
Supelco
Retinyl Palmitate (Vitamin A Palmitate), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Retinyl palmitate, potency: ≥1,700,000 USP units per g
Sigma-Aldrich
Retinyl palmitate, Type IV, ~1,800,000 USP units/g, oil