Merck
  • Effects of Devil's Claw (Harpagophytum procumbens) on the multidrug transporter ABCB1/P-glycoprotein.
Effects of Devil's Claw (Harpagophytum procumbens) on the multidrug transporter ABCB1/P-glycoprotein.
Phytomedicine : international journal of phytotherapy and phytopharmacology (2009-07-07)
Nadia Romiti, Gianfranco Tramonti, Alessandro Corti, Elisabetta Chieli
ABSTRACT
Devil's Claw (Harpagophytum procumbens) a plant native to Southern Africa, has historically been used in traditional medicine to treat a wide range of diseases and currently is widely employed as anti-inflammatory and pain-relieving natural remedy in Europe and other parts of the world. Little is known about possible herb-drug interactions arising from effects of Devil's Claw on the major drug metabolizing enzymes or transporters. This study evaluated in vitro the effects of Devil's Claw on the multidrug transporter ABCB1/P-glycoprotein. The effects of three commercially available Devil's Claw preparations and that of pure harpagoside were studied in the human kidney (HK-2) proximal tubule cell line, constitutively expressing ABCB1/P-glycoprotein (P-gp). Pgp activity and expression were tested by the calcein-AM test and by Western blotting, respectively. Commercial preparations inhibited P-gp activity, even if to a different extent, while pure harpagoside was almost ineffective. In cells cultured for three days in the presence of Devil's Claw preparations or pure harpagoside, a dose-dependent P-gp upregulation was found. Our results demonstrate for the first time that Devil's Claw may interact with the multidrug transporter ABCB1/P-gp, the effect not appearing strictly related to the harpagoside relative content. Modulation of both P-gp activity and P-gp expression by Devil's Claw raise the possibility of herb-drug interactions, to be further explored in depth.
Product Description
Supelco
Harpagoside, analytical standard
Harpagoside, primary reference standard