Merck
  • A synthetic route to 3-C-alkyl (or 3-C-phenyl-) 2,3-dideoxy-D-erythro-pentono-1,4-lactones: intermediates in the synthesis of 2(3H)-furanones.
A synthetic route to 3-C-alkyl (or 3-C-phenyl-) 2,3-dideoxy-D-erythro-pentono-1,4-lactones: intermediates in the synthesis of 2(3H)-furanones.
Carbohydrate research (1994-02-03)
P C Raveendranath, V J Blazis, K Agyei-Aye, A K Hebbler, L N Gentile, E S Hawkins, S C Johnson, D C Baker
ABSTRACT
A series of 3-C-alkyl- (and 3-C-phenyl-) 2,3-dideoxy-D-erythro-pentono-1,4-lactones, compounds which are important in the synthesis of modified nucleosides and antibiotic sugars, were synthesized from D-ribonolactone. By a route that proceeded via 5-O-protected D-ribonolactone, 5-O-protected 2,3-dideoxy-D-glycero-pent-2-enono-1,4-lactones were synthesized and reacted with R2CuLi or a complex PhSCu(RMgBr)n to give respectively the 3-C-alkyl or 3-C-phenyl compounds. Details of the preparation of the O-protected intermediates, as well as the selection of the organometallic reagents, are provided.
Sigma-Aldrich
D-(+)-Ribonic γ-lactone, 97%