Merck
  • 4-[1-(Substituted aryl/alkyl carbonyl)-benzoimidazol-2-yl]-benzenesulfonic acids: synthesis, antimicrobial activity, QSAR studies, and antiviral evaluation.
4-[1-(Substituted aryl/alkyl carbonyl)-benzoimidazol-2-yl]-benzenesulfonic acids: synthesis, antimicrobial activity, QSAR studies, and antiviral evaluation.
European journal of medicinal chemistry (2010-10-26)
Snehlata Yadav, Pradeep Kumar, Erik De Clercq, Jan Balzarini, Christophe Pannecouque, Sharwan Kumar Dewan, Balasubramanian Narasimhan
ABSTRACT
A series of 4-[1-(substituted aryl/alkyl carbonyl)-benzoimidazol-2-yl]-benzene sulphonic acids (1-20) was synthesized and evaluated, in vitro, for their antimicrobial activity and the results indicated that compounds 4-[1-(4-Nitrobenzoyl)-1H-benzoimidazol-2-yl]-benzenesulfonic acid (9) and 4-(1-octadec-9-enoyl-1H-benzoimidazol-2-yl)-benzenesulfonic acid (18) were found to be the most active ones. QSAR investigations indicated that the multi-target QSAR model was effective in describing the antimicrobial activity over the one-target QSAR models. Further the mt-QSAR model indicated the importance of the topological parameter, Balaban index (J) followed by the electronic parameter, LUMO and topological parameter, valence second order molecular connectivity index (2χv) in describing the antimicrobial activity of synthesized compounds (1-20).
Sigma-Aldrich
Benzenesulfonic acid, technical grade, 90%
Sigma-Aldrich
Benzenesulfonic acid, 98.0% (T)
Sigma-Aldrich
Sodium benzenesulfonate, 97%