Merck
  • The histone H3/H4 chaperone CHAF1B prevents the mislocalization of CENP-A for chromosomal stability.
The histone H3/H4 chaperone CHAF1B prevents the mislocalization of CENP-A for chromosomal stability.
Journal of cell science (2023-05-02)
Roshan L Shrestha, Vinutha Balachandra, Jee Hun Kim, Austin Rossi, Pranathi Vadlamani, Subhash Chandra Sethi, Laurent Ozbun, Shinjen Lin, Ken Chin-Chien Cheng, Raj Chari, Tatiana S Karpova, Gianluca Pegoraro, Daniel R Foltz, Natasha J Caplen, Munira A Basrai
ABSTRACT
Restricting the localization of the evolutionarily conserved centromeric histone H3 variant CENP-A to centromeres prevents chromosomal instability (CIN). The mislocalization of CENP-A to non-centromeric regions contributes to CIN in yeasts, flies and human cells. Even though overexpression and mislocalization of CENP-A have been reported in cancers, the mechanisms responsible for its mislocalization remain poorly understood. Here, we used an imaging-based high-throughput RNAi screen to identify factors that prevent mislocalization of overexpressed YFP-tagged CENP-A (YFP-CENP-A) in HeLa cells. Among the top five candidates in the screen - the depletion of which showed increased nuclear YFP-CENP-A fluorescence - were the histone chaperones CHAF1B (or p60) and CHAF1A (or p150). Follow-up validation and characterization experiments showed that CHAF1B-depleted cells exhibited CENP-A mislocalization, CIN phenotypes and increased enrichment of CENP-A in chromatin fractions. The depletion of DAXX, a histone H3.3 chaperone, suppressed CENP-A mislocalization and CIN in CHAF1B-depleted cells. We propose that in CHAF1B-depleted cells, DAXX promotes mislocalization of the overexpressed CENP-A to non-centromeric regions, resulting in CIN. In summary, we identified regulators of CENP-A localization and defined a role for CHAF1B in preventing DAXX-dependent CENP-A mislocalization and CIN.
Sigma-Aldrich
Anti-HJURP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-CENP-A Antibody, Upstate®, from rabbit