Merck
  • Investigating the efficacy and safety of mineral smectite granules on wound healing.
Investigating the efficacy and safety of mineral smectite granules on wound healing.
Experimental and therapeutic medicine (2021-01-19)
Jiankun Wang, Min Wang, Lili Zhao, Li Liu, Xiang Wang, Zhining Fan
ABSTRACT
Mineral smectite granules have traditionally been used for diarrhea and gastrointestinal bleeding; however, the wound healing ability of mineral smectite granules has not been investigated in detail. Therefore, the present study explored the efficacy and safety of mineral smectite granules on wound healing. An excision wound rat model was established. The experimental group was treated with smectite granules, the positive control group was treated with starch and the blank control group was left untreated. Wound closure rates, wound healing times and histopathological analysis were compared among the three groups. The mechanism underlying the effects of mineral smectite granules on wound healing was assessed by performing picrosirius red staining and immunohistochemical staining and measuring hydroxyproline content. Based on wound closure rates, wound healing times and histopathological analysis, the results indicated that rats treated with smectite granules displayed increased wound healing efficacy compared with the other two groups. Investigation of the underlying mechanism suggested that mineral smectite granules could promote wound healing by inducing collagen fiber synthesis and increasing the number of neovessels. The present study indicated the efficacy and safety of mineral smectite granules on wound healing and provided a theoretical basis for its application in a clinical setting.
Sigma-Aldrich
Anti-Mouse IgG (Fc specific)–Biotin antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-PECAM1 antibody produced in mouse, clone 1D2-1A5, purified immunoglobulin, buffered aqueous solution