Merck
  • Simultaneous quantification of tizoxanide and tizoxanide glucuronide in mouse plasma by liquid chromatography-tandem mass spectrometry.
Simultaneous quantification of tizoxanide and tizoxanide glucuronide in mouse plasma by liquid chromatography-tandem mass spectrometry.
Biomedical chromatography : BMC (2016-04-24)
Jinqiu Liu, Fanhua Meng, Zhaofeng Li, Lin Yu, Shuangqing Peng, Jifen Guo, Haiyan Xu
ABSTRACT
Nitazoxanide (NTZ) is a broad-spectrum antimicrobial agent. Tizoxanide (T) and tizoxanide glucuronide (TG) are the major circulating metabolites after oral administration of NTZ. A rapid and specific LC-MS/MS method for the simultaneous quantification of T and TG in mouse plasma was developed and validated. A simple acetonitrile-induced protein precipitation method was employed to extract two analytes and the internal standard glipizide from 50 μL of mouse plasma. The purified samples were resolved using a C18 column with a mobile phase consisting of acetonitrile and 5 mm ammonium formate buffer (containing 0.05% formic acid) following a gradient elution. An API 3000 triple quadrupole mass spectrometer was operated under multiple reaction-monitoring mode with electrospray ionization. The precursor-to-product ion transitions m/z 264 → m/z 217 for T and m/z 440 → m/z 264 for TG were used for quantification. The developed method was linear in the concentration ranges of 1.0-500.0 ng/mL for T and 5.0-1000.0 ng/mL for TG. The intra- and inter-day precision and accuracy of the quality control samples at low, medium and high concentrations exhibited an RSD of <13.2% and the accuracy values ranged from -9.6 to 9.3%. We used this validated method to study the pharmacokinetics of T and TG in mice following oral administration of NTZ. Copyright © 2016 John Wiley & Sons, Ltd.
Sigma-Aldrich
Tizoxanide, ≥98% (HPLC)