Merck
  • High-dose chemotherapy and CD34-selected peripheral blood progenitor cell transplantation for patients with breast cancer metastatic to bone and/or bone marrow.
High-dose chemotherapy and CD34-selected peripheral blood progenitor cell transplantation for patients with breast cancer metastatic to bone and/or bone marrow.
Bone marrow transplantation (2002-01-10)
J L Klein, C Hamm, R D Dansey, C Karanes, E Abella, L Cassells, W P Peters, R D Baynes
ABSTRACT
Fifty women with breast cancer metastatic to bone or bone marrow involvement on light microscopy at the time of initial evaluation were treated with high-dose chemotherapy (HDC) and peripheral blood progenitor cell (PBPC) transplantation with CD34(+) cell selection using the Isolex 300i system. All patients received induction chemotherapy. PBPC were mobilized with chemotherapy and granulocyte colony-stimulating factor. The median CD34(+) progenitor purity was 94.7% (range 72-98.7%) and recovery 38.4% (range 21-60%). Forty-eight hours after HDC with cyclophosphamide, cisplatin and carmustine, PBPC were reinfused. Median time to neutrophil count >0.5 x 10(9)/l was 9 (range 9-12) days and to platelet transfusion independence 11 (4-30) days. These data demonstrate that selected CD34(+) PBPCs allow rapid hematologic reconstitution after HDC. During follow-up, 23% of patients developed herpes zoster. Two patients developed cytomegalovirus infections. Three patients developed fungal infections. The development of these infections was not associated with steroid use but appeared more frequently in patients with diabetes mellitus. Seventy-four per cent of patients received steroids for pulmonary toxicity. Treatment-related mortality was 4%. Progression-free survival and overall survival at 35 months was 22.4% and 40.5%, with a median of 11.4 months and 15.4 months, respectively.
Sigma-Aldrich
Alkaline Phosphatase-Anti-Alkaline Phosphatase antibody produced in mouse, clone AP1B9, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-Cytokeratin, pan (Mixture) antibody produced in mouse, clone C-11+PCK-26+CY-90+KS-1A3+M20+A53-B/A2, ascites fluid