Merck
  • Sterol Structure Strongly Modulates Membrane-Islet Amyloid Polypeptide Interactions.
Sterol Structure Strongly Modulates Membrane-Islet Amyloid Polypeptide Interactions.
Biochemistry (2018-01-27)
Xiaoxue Zhang, Erwin London, Daniel P Raleigh
ABSTRACT
Amyloid formation has been implicated in a wide range of human diseases, and the interaction of amyloidogenic proteins with membranes are believed to be important for many of them. In type-2 diabetes, human islet amyloid polypeptide (IAPP) forms amyloids, which contribute to β-cell death and dysfunction in the disease. IAPP-membrane interactions are potential mechanisms of cytotoxicity. In vitro studies have shown that cholesterol significantly modulates the ability of model membranes to induce IAPP amyloid formation and IAPP-mediated membrane damage. It is not known if this is due to the general effects of cholesterol on membranes or because of specific sterol-IAPP interactions. The effects of replacing cholesterol with eight other sterols/steroids on IAPP binding to model membranes, membrane disruption, and membrane-mediated amyloid formation were examined. The primary effect of the sterols/steroids on the IAPP-membrane interactions was found to reflect their effect upon membrane order as judged by fluorescence anisotropy measurements. Specific IAPP-sterol/steroid interactions have smaller effects. The fraction of vesicles that bind IAPP was inversely correlated with the sterols/steroids' effect on membrane order, as was the extent of IAPP-induced membrane leakage and the time to form amyloids. The correlation between the fraction of vesicles binding IAPP and membrane leakage was particularly tight, suggesting the restriction of IAPP to a subset of vesicles is responsible for incomplete leakage.
Sigma-Aldrich
5α-Cholestan-3-one, crystalline
Sigma-Aldrich
DPH, ≥98% (HPLC)
Sigma-Aldrich
5-Pregnen-3β-ol-20-one, ≥98%