
Valsartan for peak identification

Synonym(s):
Valsartan, N-(1-Oxopentyl)-N-[[2′-(2H-tetrazol-5-yl)[1,1′-biphenyl]-4-yl]methyl]-L-valine
Empirical Formula (Hill Notation):
C24H29N5O3
CAS Number:
Molecular Weight:
435.52
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing