
Tarenflurbil

Synonym(s):
(R)-(−)-2-Fluoro-α-methyl-4-biphenylacetic acid, (R)-2-Flurbiprofen, Tarenflurbil
Linear Formula:
C6H5C6H3(F)CH(CH3)CO2H
CAS Number:
Molecular Weight:
244.26
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing