
Sulfadimethoxine for peak identification

Synonym(s):
Sulfadimethoxine, 4-Amino-N-(2,6-dimethoxy-4-pyrimidinyl)benzenesulfonamide
Empirical Formula (Hill Notation):
C12H14N4O4S
CAS Number:
Molecular Weight:
310.33
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing