
Sulbactam Related Compound E / EP Impurity E

Synonym(s):
(2S,5R)-6,6-Dibromo-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid 4,4-dioxide, 6,6-Dibromopenicillanic acid 1,1-dioxide
Empirical Formula (Hill Notation):
C8H9Br2NO5S
CAS Number:
Molecular Weight:
391.03
Products
Brand
Formula Weight
Agency
Quality Segment
Application
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing