![(S)-4-Methyl-2-oxazolidinone](/deepweb/assets/sigmaaldrich/product/structures/702/307/46466e66-5ece-4940-9f73-6d9120922974/640/46466e66-5ece-4940-9f73-6d9120922974.png)
(S)-4-Methyl-2-oxazolidinone
![(S)-4-Methyl-2-oxazolidinone](/deepweb/assets/sigmaaldrich/product/structures/702/307/46466e66-5ece-4940-9f73-6d9120922974/640/46466e66-5ece-4940-9f73-6d9120922974.png)
Empirical Formula (Hill Notation):
C4H7NO2
CAS Number:
Molecular Weight:
101.10
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing