
Racecadotril impurity A

Synonym(s):
Thioacetic acid, Thiacetic acid
Linear Formula:
CH3COSH
CAS Number:
Molecular Weight:
76.12
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing