
Quetiapine hemifumarate salt

Synonym(s):
2-[2-(4-Dibenzo[b,f][1,4]thiazepin-11-yl-1-piperazinyl)ethoxy]ethanol hemifumarate, Quetiapine fumarate
Empirical Formula (Hill Notation):
C21H25N3O2S · 0.5C4H4O4
CAS Number:
Molecular Weight:
441.54
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing